Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-3C7]

Supplier: Novus Biologicals
NBP1-18970 NBP1-18970SS
NOVUNBP1-18970EA 554 EUR
NOVUNBP1-18970 NOVUNBP1-18970SS
Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-3C7]
Antibodies
Type: Primary
Antigen: Oval Cell Marker
Clonality: monoclonal
Clone: OC2-3C7
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR