Anti-LDHA Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-48336 NBP1-48336SS
NOVUNBP1-48336EA 638 EUR
NOVUNBP1-48336 NOVUNBP1-48336SS
Anti-LDHA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Lactate Dehydrogenase A Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Bovine, Porcine.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100

Type: Primary
Antigen: LDHA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR