Anti-TET1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-78966 NBP1-78966SS
NOVUNBP1-78966EA 554 EUR
NOVUNBP1-78966 NOVUNBP1-78966SS
Anti-TET1 Rabbit Polyclonal Antibody
Antibodies
The TET1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TET1. This antibody reacts with mouse. The TET1 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Western Blot (Negative), ICC / IF (Negative).

Recommended Dilutions: Immunohistochemistry (Parafin): 1:400

Type: Primary
Antigen: TET1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol TET1
Antigen name Tet Methylcytosine Dioxygenase 1
Environmentally Preferable
Antigen synonyms CXXC-type zinc finger protein 6|ten-eleven translocation 1 gene protein|bA119F7.1|methylcytosine dioxygenase TET1|CXXC6|tet oncogene 1|leukemia-associated protein with a CXXC domain|ten-eleven translocation-1|LCX|CXXC finger 6|CXXC zinc finger 6
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Mouse
Host Rabbit
Gene ID 80312
Isotype
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR