Anti-Fibronectin Mouse monoclonal antibody unconjugated [clone: 2F4]
Supplier: Novus Biologicals
NBP2-22113 NBP2-22113SS
NOVUNBP2-22113EA 586 EUR
NOVUNBP2-22113 NOVUNBP2-22113SS
Anti-Fibronectin Mouse monoclonal antibody unconjugated [clone: 2F4]
Antibodies
Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: monoclonal
Clone: 2F4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human