Anti-CCL2 Mouse monoclonal antibody unconjugated [clone: 2D8]

Supplier: Novus Biologicals
NBP2-22115 NBP2-22115SS
NOVUNBP2-22115EA 732 EUR
NOVUNBP2-22115 NOVUNBP2-22115SS
Anti-CCL2 Mouse monoclonal antibody unconjugated [clone: 2D8]
Antibodies
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: monoclonal
Clone: 2D8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR