Anti-PDX1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-22150 NBP2-22150SS
NOVUNBP2-22150EA 558 EUR
NOVUNBP2-22150 NOVUNBP2-22150SS
Anti-PDX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PDX1 (pancreatic and duodenal homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR