Anti-Bat Immunoglobulin Mouse monoclonal antibody unconjugated [clone: BT1-4F10]

Supplier: Novus Biologicals
NBP2-23483 NBP2-23483SS
NOVUNBP2-23483EA 567 EUR
NOVUNBP2-23483 NOVUNBP2-23483SS
Anti-Bat Immunoglobulin Mouse monoclonal antibody unconjugated [clone: BT1-4F10]
Antibodies
Type: Primary
Antigen: Bat Immunoglobulin
Clonality: monoclonal
Clone: BT1-4F10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR