Anti-PON1 Mouse Monoclonal Antibody [clone: 4G8D3]

Supplier: Novus Biologicals
NBP2-23610 NBP2-23610SS
NOVUNBP2-23610EA 554 EUR
NOVUNBP2-23610 NOVUNBP2-23610SS
Anti-PON1 Mouse Monoclonal Antibody [clone: 4G8D3]
Antibodies
Type: Primary
Antigen: PON1 (paraoxonase 1)
Clonality: monoclonal
Clone: 4G8D3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR