Anti-EIF2A Mouse monoclonal antibody unconjugated [clone: 3A7B11]

Supplier: Novus Biologicals
NBP2-26296 NBP2-26296SS
NOVUNBP2-26296EA 554 EUR
NOVUNBP2-26296 NOVUNBP2-26296SS
Anti-EIF2A Mouse monoclonal antibody unconjugated [clone: 3A7B11]
Antibodies
Type: Primary
Antigen: EIF2A (eukaryotic translation initiation factor 2A, 65kDa)
Clonality: monoclonal
Clone: 3A7B11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR