Anti-CXCR4 Chicken Polyclonal Antibody
Supplier: US Biological
C8350-05
USBIC8350-05EA 690 EUR
USBIC8350-05
Anti-CXCR4 Chicken Polyclonal Antibody
Antibodies
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Bovine, Human, Mouse, Rat