Filter tips, Mµlti Low Retention
Supplier: Sorenson Bioscience
35240 35260 35200 35210 35220
732-3253EA 159 EUR
732-3253 732-3254 732-3249 732-3250 732-3251
Filter tips, Mµlti Low Retention
Pipette Tips
The Mμlti Low Retention filter tip range, with its special bonded polymer technology, reduces the adhesion of molecules to the filter tip, and thereby sample loss up to 10 fold. The tips are recommended for all experiments where both contamination free pipetting and a high quantitative sample recovery is essential.
  • Bonded polymer technology for a particularly 'smooth' surface to minimise sample loss
  • Hydrophobic and chemically inert filter to protect against aerosol and liquid contamination
  • Certified free from RNases, DNases, human DNA, PCR inhibitors, pyrogens and heavy metals
  • Firm fixation but easy removal on the widest range of pipettes
  • Sterilised by γ-irradiation and also autoclavable
Caution: * Not suitable for use with Eppendorf 2,5 μl pipettes