HPLC columns, OmniSpher™ C18
Supplier: VARIAN
CP27831 CP27806 CP27813 CP27833 CP27835 CP27836
VARICP27831EA 1020 EUR
VARICP27831 VARICP27806 VARICP27813 VARICP27833 VARICP27835 VARICP27836
HPLC columns, OmniSpher™ C18
Chromatography Columns
OmniSpher™ is an ultra-high bonded silica designed for the analysis of mixtures containing either acidic or basic functionality that would normally tail on more traditional columns.
  • Ultra-high purity silica backbone
  • Excellent column stability
  • Easy to scale up