Ice flakers, MF series
Supplier: SCOTSMAN
MF26/SB193 MF36 MF26
SCOMMF26/SB193EA 5750 EUR
SCOMMF26/SB193 SCOMMF36 SCOMMF26
Ice flakers, MF series
Ice Makers
Modular ice flakers is designed to produce hygienic, clear, pure ice ideal for medical and laboratory applications. It requires both a water and electricity supply and air or water-cooled condenser. Stainless steel finish, with ABS plastic lid.
  • Reduced-maintenance
  • Self-lubricating gear-reducer
  • Oversized hermetic compressor
  • Removable side panels on all four sides to aid routine maintenance and servicing
Delivery information: All units are delivered without a plug.