Preparative HPLC cartridges
Supplier: Agilent
870050-902 870050-906 870100-902 870100-906 870150-902 870150-906 970050-902 970050-906 970100-902 970100-906 970150-902 970150-906 770150-902 820212-930 897250-102 820212-921 770050-902 770100-902 820212-914 820212-915 820212-918 820212-920 820212-924 820212-925 820212-926 820212-928 820212-933 868050-901 868100-901 868150-901 870050-914 870100-914 870150-914 877150-102 877150-106 877150-114 877250-101 877250-102 877250-105 877250-106 877250-112 877250-114 877952-101 877952-102 877952-105 877952-106 877952-108 877967-102 878150-101 878250-101 895050-902 895050-906 895050-909 895100-902 895100-906 895100-909 895150-902 895150-906 895150-909 897150-102 897150-106 897150-109 897250-105 897250-106 897250-109 977150-102 977150-106 977250-102 977250-106
HEWL870050-902EA 0 EUR
HEWL870050-902 HEWL870050-906 HEWL870100-902 HEWL870100-906 HEWL870150-902 HEWL870150-906 HEWL970050-902 HEWL970050-906 HEWL970100-902 HEWL970100-906 HEWL970150-902 HEWL970150-906 HEWL770150-902 HEWL820212-930 HEWL897250-102 HEWL820212-921 HEWL770050-902 HEWL770100-902 HEWL820212-914 HEWL820212-915 HEWL820212-918 HEWL820212-920 HEWL820212-924 HEWL820212-925 HEWL820212-926 HEWL820212-928 HEWL820212-933 HEWL868050-901 HEWL868100-901 HEWL868150-901 HEWL870050-914 HEWL870100-914 HEWL870150-914 HEWL877150-102 HEWL877150-106 HEWL877150-114 HEWL877250-101 HEWL877250-102 HEWL877250-105 HEWL877250-106 HEWL877250-112 HEWL877250-114 HEWL877952-101 HEWL877952-102 HEWL877952-105 HEWL877952-106 HEWL877952-108 HEWL877967-102 HEWL878150-101 HEWL878250-101 HEWL895050-902 HEWL895050-906 HEWL895050-909 HEWL895100-902 HEWL895100-906 HEWL895100-909 HEWL895150-902 HEWL895150-906 HEWL895150-909 HEWL897150-102 HEWL897150-106 HEWL897150-109 HEWL897250-105 HEWL897250-106 HEWL897250-109 HEWL977150-102 HEWL977150-106 HEWL977250-102 HEWL977250-106
Preparative HPLC cartridges
Chromatography Columns
These cartridge columns are with reusable end fittings that have a unique design which makes them easy to install with finger-tight connections.
  • Available in a variety of bonded phases
  • Easy scale up from analytical to preparative scale without losing resolution
  • Fast preparative separation