Separating funnels
Supplier: QVF LABORTECHNIK
ALG0050703 ALG0050704 ALG0050706 ALG0050702 ALG0050701
QVFLALG0050703EA 129 EUR
QVFLALG0050703 QVFLALG0050704 QVFLALG0050706 QVFLALG0050702 QVFLALG0050701
Separating funnels
Funnels Separating Funnels
Made of borosilicate glass.
  • Stopcock with glass key
  • Without graduations
  • Excellent chemical and temperature resistance
The separating funnels are used for discontinuous shaking out of solvents.
Certifications: Complies to DIN 12 451.