Connectors, closed, Stronghold™
Supplier: HARVARD APPARATUS
53-2012-W 53-2012-R
241-2211EA 20.7 EUR
241-2211 241-2212
Connectors, closed, Stronghold™
Clamps General Purpose Clamps
The most common use of this closed connector is to firmly mount the stem of a clamp horizontally on a vertical lattice rod or stand. It holds rods or clamp stems up to 13 mm Ø. The two rods are held at a right angle to each other.
  • Precision injection molded nylon with 30% glass fiber reinforcement provides enormous strength at a great saving in weight
  • There is positively no corrosion possible
  • Material is perfect for laboratory use since it has the very high melting point of 221 °C and good resistance to chemicals, particularly salt solutions and most