Anti-CXCL16 Goat Polyclonal Antibody

Supplier: US Biological
C8298-04A-100 C8298-04A-50
USBIC8298-04A-100EA 647 EUR
USBIC8298-04A-100 USBIC8298-04A-50
Anti-CXCL16 Goat Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 0.5-2.0 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: CXCL16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR