Anti-Osteoinductive Factor Rabbit Polyclonal Antibody
Supplier: US Biological
O8062-27-20 O8062-27-100
USBIO8062-27-20EA 379 EUR
USBIO8062-27-20 USBIO8062-27-100
Anti-Osteoinductive Factor Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:5000
Type: Primary
Antigen: Osteoinductive Factor
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,