Anti-Signal Transducer And Activator Of Transcription 3 Rabbit Polyclonal Antibody
Supplier: US Biological
142289 142289-100
USBI142289EA 475 EUR
USBI142289 USBI142289-100
Anti-Signal Transducer And Activator Of Transcription 3 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:100-200; Western Blot: 1:100-400; Immunochemistry: 1:100-500
Type: Primary
Antigen: Signal Transducer And Activator Of Transcription 3
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat,