Anti-Calprotectin Mouse Monoclonal Antibody
Supplier: US Biological
143123 143123-100
USBI143123EA 391 EUR
USBI143123 USBI143123-100
Anti-Calprotectin Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Calprotectin
Clonality: Monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now