Anti-Trail Rabbit Polyclonal Antibody (Biotin)

Supplier: US Biological
143686 143686-50
USBI143686EA 324 EUR
USBI143686 USBI143686-50
Anti-Trail Rabbit Polyclonal Antibody (Biotin)
Antibodies
Recommended dilution: ELISA: 0.25-1 ?g/ml

Type: Primary
Antigen: Trail
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR