Millex® Syringe Filters, PTFE Membrane
Supplier: Merck
SLFG65010 SLFG55010 SLFG05000 SLFG02550 SLFGL25BS SLFGN25VS SLFG025LS SLFA05000 SLFA05010 SLFG0250M SLFG65000 SLFG75000 SLFG75010 SLFH05000 SLFH05010 SLFG05010
MILFSLFG65010EA 186 EUR
MILFSLFG65010 516-6551 MILFSLFG05000 514-0409 MILFSLFGL25BS MILFSLFGN25VS 514-0410 MILFSLFA05000 MILFSLFA05010 MILFSLFG0250M MILFSLFG65000 MILFSLFG75000 MILFSLFG75010 MILFSLFH05000 MILFSLFH05010 MILFSLFG05010
Millex® Syringe Filters, PTFE Membrane
Filters Syringe Filters
Syringe filters with hydrophobic PTFE membrane. Used in vacuum line protection, transducer protection and gas filtration. Available as either 25 mm or pump-driven Millex®-FG₅₀ 50 mm diameter filters.
Filters are either non sterile or sterilised with ethylene oxide.
Order Now
SPECIFICATIONS
Environmentally Preferable
Max. operating pressure 4,1 bar 4,1 bar 2,0 bar 5,2 bar
Max. temperature 130 °C 121 °C 45 °C 45 °C