Hanks balanced salt solution (HBSS)
04-315Q
LONZ04-315QEA 45.8 EUR
LONZ04-315Q
Hanks balanced salt solution (HBSS)
Buffers Cell Culture Buffers
Hanks balanced salt solution is used in a wide variety of tissue culture applications like to maintain a physiological pH for cells maintained in non CO₂ conditions, diluting cells before counting, washing cells before dissociating and transporting cells or tissues.