DNA ladder
Supplier: LONZA
50322
50323
50330
50321
LONZ50322EA
237
EUR
LONZ50322
LONZ50323
LONZ50330
LONZ50321
DNA ladder
Electrophoresis Reagents
Molecular Weight Markers
Thse DNA ladders provides rapid and easy size determination of the DNA samples. Each band has an ideal 50 % G-C content for precise sizing and sharp resolution. It is free from contaminating plasmid DNA with minimal background.