DNA markers
Supplier: LONZA
50461
50471
LONZ50461EA
362
EUR
LONZ50461
LONZ50471
DNA markers
Electrophoresis Reagents
Molecular Weight Markers
DNA marker is for rapid size estimation of PCR products and restriction fragments. DNA can be visualised by ethidium bromide staining or kinased with radiolabelled ³²P for detection by autoradiography.