Megabase DNA standards
50411
LONZ50411EA
0
EUR
LONZ50411
Megabase DNA standards
Electrophoresis Reagents
Molecular Weight Markers
Megabase DNA standards are specially prepared in InCert™ Agarose for use in pulsed field gel electrophoresis (PFGE).
- Minimal lot-to-lot variation
- Each lot is performance tested for reliable results