Gas detection tubes for sulphur dioxide
Supplier: Dräger
6727101 CH24201 CH31701 6728491 8101531
151-0146EA 87.75 EUR
151-0146 151-0194 151-0207 151-0154 118-0308
Gas detection tubes for sulphur dioxide
Gas Tubes
To measure the 'spot concentrations' of sulphur dioxide for use with an Accuro® pump.
Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.
The tube sulphur dioxide1/a has a prelayer so that H₂S in the TLV range is retained and does not interfere. Hydrogen sulphide and acidic gases may interfere with other tubes.
One stroke is equivalent to a volume of 100 ±5 ml of gas passing through the tube.
The standard measuringMore Product Information
Order Now
SPECIFICATIONS
Model Sulphur dioxide 0.1/a Sulphur dioxide 20/a Sulphur dioxide 1/a Sulphur dioxide 0.5/a Sulphur dioxide 50/b
Environmentally Preferable
Colour change Yellow to orange Brown yellow to white Grey blue to white Grey blue to white Blue to yellow
Humidity 3 -15 mg/l H₂O ≤ 30 mg/l H₂O 3 -20 mg/l H₂O ≤ 20 mg/l H₂O 1 -15 mg/l H₂O
Measurement time 20 minutes 3 minutes 3 minutes 3/6 minutes 15 seconds/3 minutes
No. of strokes 100 10 10 10/20 1/10
Standard deviation ±10 - 15 % ±10 - 15 % ±10 - 15 % ±10 - 15 % ±10 - 15 %
Standard range 0,1 - 3 ppm 20 - 200 ppm 1 - 25 ppm 1 - 25/0,5 - 5 ppm 400 - 8000 / 50 - 500 ppm
Temperature range 10…30 °C 0…40 °C 15…25 °C 15…30 °C 0…50 °C