Septa, Merlin Microseal™
Supplier: Restek
22812 22810 22809 22811 22813 22814 22815
REST22812EA 253 EUR
REST22812 REST22810 REST22809 REST22811 REST22813 REST22814 REST22815
Septa, Merlin Microseal™
Septa
Microseal septum incorporates two separate sealing mechanisms. These sliding seals eliminate septum coring and the resulting accumulation of septum crumbs in the injection port liner. The septum uses a 23-gauge (0,63 mm, 0,025”) needle or probe with a blunt, truncated conical tip. Since the syringe plunger end details are determined by manual or autosampler compatibility. The advantages of the septum include elimination of septum coring, longer life, consistent and low needle insertion force.
  • Allow operation from 206,8 to 6894,7 mbar (general-purpose microseal septa) or 68,9 to 3102,6 mbar (low-pressure microseal septa)
  • 400 °C maximum injection port temperature