Capillary columns, Rtx®-5MS (fused silica)
Supplier: Restek
12605 12606 12608 12609 12611 12612 12620 12621 12623 12624 12626 12627 12635 12637 12638 12639 12640 12641 12642 12650 12651 12652 12653 12654 12655 12667 12670 12690 12691 12692 12693 12694 12695 12605-123 12608-600 12620-364 12620-600 12623-042 12623-106600 12623-112 12623-124042 12623-303 12623-305 12623-600 12624-042 12624-6850 12626-118 12626-600 12626-6850 12638-101 12638-337 12638-600 12639-106 12639-600 12641-6850 12650-104 12650-123 12655-6850 12694-304
REST12605EA 499 EUR
REST12605 REST12606 REST12608 REST12609 REST12611 REST12612 REST12620 REST12621 REST12623 REST12624 REST12626 REST12627 REST12635 REST12637 REST12638 REST12639 REST12640 REST12641 REST12642 REST12650 REST12651 REST12652 REST12653 REST12654 REST12655 REST12667 REST12670 REST12690 REST12691 REST12692 REST12693 REST12694 REST12695 REST12605-123 REST12608-600 REST12620-364 REST12620-600 REST12623-042 REST12623-106600 REST12623-112 REST12623-124042 REST12623-303 REST12623-305 REST12623-600 REST12624-042 REST12624-6850 REST12626-118 REST12626-600 REST12626-6850 REST12638-101 REST12638-337 REST12638-600 REST12639-106 REST12639-600 REST12641-6850 REST12650-104 REST12650-123 REST12655-6850 REST12694-304
Capillary columns, Rtx®-5MS (fused silica)
Chromatography Columns
General purpose columns for drugs, solvent impurities, pesticides, hydrocarbons, PCB congeners (e.g. Aroclor mixes), essential oils, semivolatiles. Column specifically tested for low-bleed performance.
Low-polarity phase; Crossbond® diphenyl dimethyl polysiloxane
Temperature range: −60 to 350 ºC
Similar phases: DB-5, HP-5, HP-5MS, Ultra-2, SPB-5, CP-Sil 8 CB, ZB-5.
Equivalent to USP G27 and G36 phases.