Connector kits, SeCure® 'Y'
Supplier: Restek
20276 20277 20278 20279
REST20276EA 409 EUR
REST20276 REST20277 REST20278 REST20279
Connector kits, SeCure® 'Y'
Fittings and Connectors
Connector kits connects two analytical columns to a transfer line or guard column. They have a reliable seal integrity and will not unexpectedly disconnect during temperature programmed analyses. The open design allows visual confirmation of the seal for added confidence in the connection.
Combine the 'Y' Press-Tight® connector with the strength of a metal union. The ferrules and knurled nuts hold the fused silica tubing in place, which prevents the tubing from unexpectedly disconnecting, even at temperatures as high as 400 °C.
Delivery information: Kits include: SeCure® 'Y' connector body, three knurled nuts, universal 'Y' Press-Tight® union and three ferrules.