Connectors, Press-Tight®
Supplier: Restek
20401 20400 20430 20429 20402 20403 20404 20405 20406 20469 20485 20486 20449 20480 20405-261 20406-261 20487
REST20401EA 331 EUR
REST20401 REST20400 REST20430 REST20429 REST20402 REST20403 REST20404 REST20405 REST20406 REST20469 REST20485 REST20486 REST20449 REST20480 REST20405-261 REST20406-261 REST20487
Connectors, Press-Tight®
Fittings and Connectors
Deactivated Press-Tight® connectors maintain complete inertness along the GC flow path and Siltek®-deactivated connectors are ideal for organochlorine pesticides analysis. These connectors fit 0,33 to 0,74 mm ext. Ø and 0,1 to 0,53 int. Ø columns.
Universal Press-Tight® connector: Connects a guard column to an analytical column, repairs a broken column and also connects a column outlet to a transfer line.
Universal 'Y' Press-Tight® connector: Connectors are an alternative method of performing dual-column coni rmational analyses. Split sample flow onto two columns and perform confirmation analysis with a single injection. Split a single column flow to two detectors.
Universal angled 'Y' Press-Tight® connector: Performs confirmation analysis with a single injection. Inlet and outlet ends conform to the column curvature and alleviates column-end connection strain.