ESD safety shoes, lace-up, GX 120, Atlas
Supplier: ATLAS - the shoe company
45100_S2_35 45100_S2_36 45100_S2_37 45100_S2_38 45100_S2_39 45100_S2_40 45100_S2_41 45100_S2_42 45100_S2_43
114-6827EA 232 EUR
114-6827 114-6828 114-6829 114-6830 114-6831 114-6832 114-6833 114-6834 114-6835
ESD safety shoes, lace-up, GX 120, Atlas
Shoes and Boots
ESD safety shoes made from waterproof leather and a MPU® Light sole (3,5 mm profile, oil, acid and petrol resistant, anti-static).
  • Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
  • Removable insoles; orthopedic insoles possible
  • 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
  • Protective steel toecap (200 J)
  • Electrical contact resistance of 1×10⁵ - 3,5×10⁷ Ohm
Colour: Black/blue
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC