VWR®, Rubber Stoppers, Black
Supplier: VWR Collection
59580-262 59580-386 59580-444
VWRU59580-262EA 0 EUR
VWRU59580-262 VWRU59580-386 VWRU59580-444
VWR®, Rubber Stoppers, Black
Stoppers Rubber Stoppers
These stoppers contain high percentage of rubber, less neoprene and free sulphur. Its composition ensures resiliency and pliability over a long period of time.
  • Natural rubber
  • Black
  • Withstands laboratory conditions without hardening