VWR®, Cryo-Labels
Supplier: VWR Collection
10123-830 10123-862 10123-864 10123-868
VWRU10123-830EA 0 EUR
VWRU10123-830 VWRU10123-862 VWRU10123-864 VWRU10123-868
VWR®, Cryo-Labels
Labels
These VWR® cryogenic labels are ideal for all cryostorage, liquid nitrogen, refrigeration and ultra-low temperature freezer applications.
  • Can be placed in most incubators
  • Unique adhesive that can withstand extreme temperatures from −196 to +121 °C without the need for lamination
Laser printer label sheets are in a size of 216×279 mm.
Delivery information: Rolls of labels are delivered on a 44 mm Ø core.