Safety shoes, Velcro® fastening, Magic
Supplier: U-Power
BC30335/35 BC30335/36 BC30335/37 BC30335/38 BC30335/39 BC30335/40 BC30335/41 BC30335/42 BC30335/43 BC30335/44 BC30335/45 BC30335/46 BC30335/47 BC30335/48
114-3250EA 58.3 EUR
114-3250 114-3251 114-3252 114-3253 114-3254 114-3255 114-3256 114-3257 114-3258 114-3259 114-3260 114-3261 114-3262 114-3263
Safety shoes, Velcro® fastening, Magic
Shoes and Boots
Anti-static safety shoes made from soft suede leather and a slip-resistant and hard wearing PU sole.
  • Removable anatomical FlatFit insole
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certifications: EN 20345:2011; S1P, SRC