Cut resistant gloves, uvex C300 foam
Supplier: UVEX
6054407 6054408 6054409 6054410 6054411
112-4434EA 180 EUR
112-4434 112-4435 112-4436 112-4437 112-4438
Cut resistant gloves, uvex C300 foam
Gloves
These bamboo viscose/Dyneema®/glass/polyamide gloves feature a HPE SoftGrip-Foam coating on palm and fingers. The Bamboo TwinFlex® technology ensures maximum cut resistance, exceptional abrasion resistance and outstanding wearing comfort.
  • Medium cut protection (Cut 3)
  • Extremely comfortable thanks to uvex climazone
  • Excellent dexterity, high abrasion-resistance
  • Moisture resistant
  • Silicone-free according to imprint test
Typical applications include: Aerospace and automotive industry, construction, engineering, maintenance, metal industry, oil and gas, and transportation. Suitable for multi-touch operations at industrial monitors.
Certifications: EN 388:2016 (4X42C)
OEKO-Tex® Standard 100