Isolator gloves, AlphaTec®, EPDM+
Supplier: Ansell
85-602/11 85-600/9.5 85-600/11 85-601/9.5 85-601/11 85-602/9.5
112-4478EA 1710 EUR
112-4478 112-4475 112-4474 112-4477 112-4476 112-4479
Isolator gloves, AlphaTec®, EPDM+
Gloves
These gloves are made of white diene ethylene propylene rubber (EPDM) with a black lining for breach detection. They are resistant against hydrogen peroxide solutions and common disinfecting chemicals.
  • High-quality material, in compliance with FDA CFR21 (food contact approved)
  • Comfortable and dexterous with great tactile sensitivity
  • Resist aging from exposure to oxygen, UV rays and ozone
  • Withstand temperatures up to 130 °C for repeated sterilisation
  • Halogen-free: Suitable for disposal by incineration
Designed for repeated autoclave sterilisation (up to 50 times), reducing the need for glove replacements. Ideal for use in pharmaceuticals and electronics.
Certifications: EN ISO 374-1 Type B (AMP), EN ISO 374-5 VIRUS, EN 388 (3100A); Cat. III