Escin ≥95%, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

E1378-10G E1378-1G E1378-5G
SIALE1378-10GEA 606 EUR
SIALE1378-10G SIALE1378-1G SIALE1378-5G
Escin ≥95%, Sigma-Aldrich®
Escin
Formula: C₅₅H₈₆O₂₄
MDL Number: MFCD00076054
CAS Number: 6805-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR