Pipette tips, standard
Supplier: Sorenson Bioscience
10230 LAC10410 LAC10390 LAC10450 10130
SORE10230EA 560 EUR
SORE10230 613-3048 613-3046 613-3050 SORE10130
Pipette tips, standard
Pipette Tips
Standard tips for general pipetting applications. Designed to fit most research-grade pipettors.
  • Available with low binding surface technology for reduced sample retention
  • Certified free from RNases, DNases, human DNA, PCR inhibitors and pyrogens
  • Standardization tips are ideal for multi-channel pipettors: Thin collar requires less force for loading and ejecting tips
  • XT tips (extended length): Ideal for pipetting from deep vessels
  • OneTouch tips create a leak proof seal: Easy to seal, easy to eject
Bulk
1000 loose tips packaged in resealable bags (exception: 1000 µl XT bulk bags with 768 tips)
Racked
Sturdy racks that withstand several autoclave cycles
CellRacks hold tips steadily in fully enclosed cells, minimize static and are excellent platforms for single and multichannel pipetting
Hinged racks hold the extremely popular 1000XT (50-1250 µl) tips, and traditional 1000 µl standard tips are placed in 100-hole standard racks
e•dek – Paperboard Rack System
Easy to recycle – compatible with most paperboard recycling programs
Used racks require minimal storage space – just flatten and recycle
Manufactured from paperboard sourced from sustainable forests
Printed with soy and vegetable based inks
Less weight equals reduced energy consumption and cost for shipping
TipStation – Multichannel Pipetting Platform
Saves space: 192 tips per deck, 5 decks per stack (960 tips)
Decks designed in 2 x 96 format to match microtiter plate formats
Stable platform for loading tips
Can be used with 8 or 12 channel pipettors
Lid can be positioned to expose only 96 tips
QuickRack Environmental Packaging
Quickly loads 96 tips into empty racks
Compatible with most manufacturers’ racks
Reduces waste