Barium zirconate ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
|
Warning
|
14028.A1
14028.30
14028.A1EA
156
EUR
14028.A1
14028.30
Barium zirconate ≥99% (metals basis)
Barium zirconate
Purity: 99%
CAS No.: 12009-21-1
Molecular Formula: BaZrO3
Formula Weight: 276.53
Physical Form: 1-2 Micron Powder
Density: 5.52
Odour: Odourless
Melting Point: 2500°
MDL No.: MFCD00014189
Formula:
BaO₃Zr Melting Pt: 2500 °C Density: 5.52 Storage Temperature: Ambient |
MDL Number:
MFCD00014189 CAS Number: 12009-21-1 EINECS: 234-546-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...