Lithium chloride/potassium chloride eutectic
Supplier: Thermo Fisher Scientific
|
Warning
|
18206.18
18206.30
18206.18EA
186
EUR
18206.18
18206.30
Lithium chloride/potassium chloride eutectic
Lithium chloride/potassium chloride eutectic
CAS No.: 65567-96-6
Molecular Formula: LiCl.KCl
Physical Form: -10 Mesh
Density: 2.02
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 357°
MDL No.: MFCD00144623
Formula:
Cl₂KLi Melting Pt: 357 °C Density: 2.02 Storage Temperature: Ambient |
MDL Number:
MFCD00144623 CAS Number: 65567-96-6 |