Aluminum metaphosphate Al₂O₃ 19,4%, P₂O₅ 78,9%

Supplier: Thermo Fisher Scientific
Warning

30715.30 30715.A1 30715.D9
30715.30EA 36.8 EUR
30715.30 30715.A1 30715.D9
Aluminum metaphosphate Al₂O₃ 19,4%, P₂O₅ 78,9%
Aluminum metaphosphate

CAS No.: 32823-06-6 Molecular Formula: Al(PO3)3 Formula Weight: 263.90 Physical Form: Powder Density: 2.779 Appearance: White MDL No.: MFCD00075314

Formula: Al(PO₃)₃
Density: 2.779
Storage Temperature: Ambient
MDL Number: MFCD00075314
CAS Number: 32823-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR