Ammonia solution 50% (v/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
35574.AP 35574.K2 35574.K7
35574.APEA 69.8 EUR
35574.AP 35574.K2 35574.K7
Ammonia solution 50% (v/v) in aqueous solution
Ammonia solution
CAS No.: 1336-21-6 Molecular Formula: H5NO Formula Weight: 35.05 Physical Form: Liquid Density: 0.94 Appearance: Colourless UN No.: UN2672 MDL No.: MFCD00066650
Formula: NH₃
MW: 35.05 g/mol
Density: 0.94
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III