Copper ≥99.9% (metals basis), tubing 3.175 mm (0.125 in) OD, 1.5 mm (0.059 in) ID
Supplier: Thermo Fisher Scientific
13471.RM 13471.RN
13471.RMEA 89.2 EUR
13471.RM 13471.RN
Copper ≥99.9% (metals basis), tubing 3.175 mm (0.125 in) OD, 1.5 mm (0.059 in) ID
Copper
Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545