Platinum 10% on activated carbon wetted with ca. 50% water, powder, reduced
Supplier: Thermo Fisher Scientific
Synonyms:
Platinium
38325.06
38325.14
38325.22
38325.06EA
137
EUR
38325.06
38325.14
38325.22
Platinum 10% on activated carbon wetted with ca. 50% water, powder, reduced
Platinum
CAS No.: 7440-06-4
Physical Form: 900-1100 m²/g
Odour: Odourless
MDL No.: MFCD00011179
Ordering information: Sold on a dry weight basis. Unit weight excludes water weight
Formula:
Pt MW: 195.08 g/mol Boiling Pt: 3827 Melting Pt: 1769 Storage Temperature: Ambient |
MDL Number:
MFCD00011179 CAS Number: 7440-06-4 Merck Index: 12,07689 |