Titanium(III) sulphate 20% in 1-4% sulphuric acid
Supplier: Thermo Fisher Scientific
Danger
39664.30 39664.A1 39664.A7
39664.30EA 81.4 EUR
39664.30 39664.A1 39664.A7
Titanium(III) sulphate 20% in 1-4% sulphuric acid
Titanium(III) sulphate
CAS No.: 10343-61-0 Molecular Formula: Ti2(SO4)3 Formula Weight: 383.38 Physical Form: Liquid Density: 1.15 UN No.: UN3264 MDL No.: MFCD00036317
Formula: O₁₂S₃Ti₂
MW: 383.902 g/mol
Density: 1.15
Storage Temperature: Ambient
MDL Number: MFCD00036317
CAS Number: 10343-61-0
UN: 3264
ADR: 8 III
Merck Index: 14,9476