Neodymium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Perchloric acid neodymium salt hexahydrate
40568.NB 40568.ND
40568.NBEA 91 EUR
40568.NB 40568.ND
Neodymium(III) perchlorate 50% (w/w) in aqueous solution, Reagent Grade
Neodymium(III) perchlorate hexahydrate
CAS No.: 17522-69-9 Molecular Formula: Nd(ClO4)3.6H2O Formula Weight: 550.68 (442.59anhy) Physical Form: Liquid Odour: Odourless UN No.: UN3211 MDL No.: MFCD00198195
Formula: Nd(ClO₄)₃·6H₂O
MW: 550.68 g/mol (442.59 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00198195
CAS Number: 17522-69-9
EINECS: 236-821-8
UN: 3211
ADR: 5.1 II