Polyvinyl alcohol, M.W. ∼ 10,000-26,000 98-99%, low molecular weight, hydrolysed
Supplier: Thermo Fisher Scientific
Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®
41241.14 41241.22 41241.36
41241.14EA 23.3 EUR
41241.14 41241.22 41241.36
Polyvinyl alcohol, M.W. ∼ 10,000-26,000 98-99%, low molecular weight, hydrolysed
Polyvinyl alcohol
Purity: 98-99% CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Odour: Odourless MDL No.: MFCD00081922
Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5