Chlorhexidine gluconate 20% (w/v) in aqueous solution, not sterile
Supplier: Thermo Fisher Scientific
Synonyms: 1,1'-Hexamethylenebis[5-(4-chlorophenyl)biguanide] digluconate, Peridex, 1,1'-Hexamethylenebis[5-(p-chlorophenyl)biguanide] (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid(1:2), Chlorhexidine digluconate
41385.AC 41385.AE 41385.AP 41385.AY
41385.ACEA 41.1 EUR
41385.AC 41385.AE 41385.AP 41385.AY
Chlorhexidine gluconate 20% (w/v) in aqueous solution, not sterile
Chlorhexidine gluconate
CAS No.: 18472-51-0 Molecular Formula: C22H30Cl2N10.2C6H12O7 Formula Weight: 897.76 Physical Form: Liquid Density: 1.06 Sensitivity: Light Sensitive Appearance: Colourless MDL No.: MFCD00083599
Formula: C₂₈H₄₄Cl₂N₁₀O₇S
MW: 897.76 g/mol
Density: 1.06
Storage Temperature: Ambient
MDL Number: MFCD00083599
CAS Number: 18472-51-0
EINECS: 242-354-0