Gold plating solution ∼3.7 g/l metal content, electroless
Supplier: Thermo Fisher Scientific
|
Danger
|
42307.AK
42307.K2
42307.AKEA
296
EUR
42307.AK
42307.K2
Gold plating solution ∼3.7 g/l metal content, electroless
Gold plating solution
Physical Form: Liquid
Appearance: Colourless to pale yellow
UN No.: UN2922
MDL No.: MFCD00003436
Applications: For coating nickel or copper
Storage Temperature:
Ambient |
MDL Number:
MFCD00003436 UN: 2922 ADR: 8,III |